Learn More
Medchemexpress LLC Netilmicin sulfate | 56391-57-2 | 98.0% | C21H41N5O7.5/2H2SO4 | 100 MG
SDP
Catalog No. 5000432173
Encompass_Preferred
Netilmicin (Sch 20569) sulfate is a broad-spectrum semisynthetic aminoglycoside antibiotic. It exhibits antibacterial activity against aminoglycoside-susceptible gram-negative strains and aminoglycoside-resistant strains.
  • Exhibits antibacterial activity against a range of gram-negative and gram-resistant strains including *Escherichia coli*, *Pseudomonas aeruginosa*, *Proteus*, *Staphylococcus aureus*, *Streptococcus*, *Serratia*, and *Enterobacter*, with a MIC of 0.125-8 μg/mL.
  • Shows in vivo antibacterial activity against many clinically important bacteria in CF1 mice, particularly against strains resistant to Gentamicin and Tobramycin that produce the adenylating enzyme ANT (2'').
Catalog No. 50-004-32173 Supplier Medchemexpress LLC Supplier No. HYA0086100MG
Netilmicin (Sch 20569) sulfate is a broad-spectrum semisynthetic aminoglycoside antibiotic. It exhibits antibacterial activity against aminoglycoside-susceptible gram-negative strains and aminoglycoside-resistant strains.
  • Exhibits antibacterial activity against a range of gram-negative and gram-resistant strains including *Escherichia coli*, *Pseudomonas aeruginosa*, *Proteus*, *Staphylococcus aureus*, *Streptococcus*, *Serratia*, and *Enterobacter*, with a MIC of 0.125-8 μg/mL.
  • Shows in vivo antibacterial activity against many clinically important bacteria in CF1 mice, particularly against strains resistant to Gentamicin and Tobramycin that produce the adenylating enzyme ANT (2'').