Learn More
Medchemexpress LLC Netilmicin (sulfate) | 56391-57-2 | 98.0% | C21H41N5O7.5/2H2SO4 | 50 MG
SDP
Catalog No. 5000432105
Encompass_Preferred
Netilmicin sulfate is a broad-spectrum semisynthetic aminoglycoside antibiotic. It exhibits antibacterial activity against aminoglycoside-susceptible gram-negative strains and aminoglycoside-resistant strains such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Staphylococcus aureus*, with a MIC of 0.125-8 μg/mL. This product is identified as a laboratory chemical for substance manufacturing.
  • Broad-spectrum semisynthetic aminoglycoside antibiotic
  • Antibacterial activity against gram-negative strains
  • Effective against aminoglycoside-resistant strains
  • Stable under recommended storage conditions
  • For research use only
Catalog No. 50-004-32105 Supplier Medchemexpress LLC Supplier No. HYA008650MG
Netilmicin sulfate is a broad-spectrum semisynthetic aminoglycoside antibiotic. It exhibits antibacterial activity against aminoglycoside-susceptible gram-negative strains and aminoglycoside-resistant strains such as *Escherichia coli*, *Pseudomonas aeruginosa*, and *Staphylococcus aureus*, with a MIC of 0.125-8 μg/mL. This product is identified as a laboratory chemical for substance manufacturing.
  • Broad-spectrum semisynthetic aminoglycoside antibiotic
  • Antibacterial activity against gram-negative strains
  • Effective against aminoglycoside-resistant strains
  • Stable under recommended storage conditions
  • For research use only