Learn More
TARGETMOL CHEMICALS INC NETIVUDINE 5MG
SDP
Catalog No. 502579608
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Netivudine is a potent nucleoside reverse transcriptase inhibitor (NRTIs) a nucleoside analogue with anti-varicella zoster virus activity that can be used to treat human immunodeficiency virus (HIV) infection. Netivudine works by inhibiting reverse transcriptase which reduces viral load in the body and slows the progression of the disease through its inhibitory effect. purity: 99%
Catalog No. 50-257-9608 Supplier TARGETMOL CHEMICALS INC Supplier No. T605405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Netivudine is a potent nucleoside reverse transcriptase inhibitor (NRTIs) a nucleoside analogue with anti-varicella zoster virus activity that can be used to treat human immunodeficiency virus (HIV) infection. Netivudine works by inhibiting reverse transcriptase which reduces viral load in the body and slows the progression of the disease through its inhibitory effect. purity: 99%