Learn More
Aviva Systems Biology Corp NETO1 Antibody - C-terminal region (ARP59810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NETO1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human NETO1. Alternate names: BCTL1, BTCL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-836