Learn More
Aviva Systems Biology Corp NETO2 Antibody - N-terminal region (ARP47391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NETO2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NETO2. Alternate names: FLJ10430, FLJ14724, FLJ90456, NEOT2, BTCL2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59538