Learn More
AdipoGen anti-Netrin-1 (human), mAb (rec.) (H4) (preservative-free) (500 µg)
SDP
Supplier:  AdipoGen AG27B0020PFC500
Encompass_Preferred
Recombinant Antibody. Recognizes human and mouse Netrin-1. Does not recognize Netrin-4. Species cross-reactivity: Human, Mouse. Clone: H4. Isotype: Human IgG2lambda. Applications: ELISA, FUNC. Host: HEK 293 cells. Liquid. In PBS containing 10% glycerol. Netrin-1 controls guidance of CNS commissural axons and peripheral motor axons. Its association with either DCC or some UNC5 receptors will lead to axon attraction or repulsion, respectively. It also serves as a survival factor via its association with its receptors which prevent the initiation of apoptosis. Netrin-1 is also involved in tumorigenesis by regulating apoptosis. Netrin-1 promotes atherosclerosis by retaining macrophages in the artery wall. Netrin-1 also governs induced pluripotent stem cell (iPS) formation and improves reprogramming efficiency of human and mouse somatic cells by limiting apoptosis mediated by Netrin-1 receptors DCC or UNC5b.
Catalog No. 50-205-3044