Learn More
AdipoGen anti-Netrin-4, mAb (Nely-1) (100 µg)
SDP
Supplier:  AdipoGen AG20B0039C100
Encompass_Preferred
Monoclonal Antibody. Recognizes human and mouse Netrin-4 Isotype: Mouse IgG1kappa. Clone: Nely-1. Applications: ELISA, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Netrin-4 is a 628 amino acid laminin-related secreted protein that promotes neurite elongation at low concentrations but inhibits neurite extension at high concentrations. Recent evidence suggest that Netrin family members act as regulators of both neuronal and vascular growth and are implicated in controling epithelial cell branching morphogenesis in the pancreas, breast and lung. Netrin-4 inhibits proliferation of a variety of human tumor cells. Netrin-4 also induces proliferation, migration, adhesion, tube formation and survival of human lymphatic endothelial cells and seems to function as a lymphangiogenic factor contributing to tumor dissemination.
Catalog No. 50-205-2947