Learn More
Medchemexpress LLC Netrin receptor DCC/ 100ug | 100ug
SDP
Catalog No. 5000194214
Encompass_Preferred
The netrin receptor DCC guides axonal attraction in neuronal growth cones during nervous system development Interaction with UNC5 protein induces axonal repulsion Netrin receptor DCC/DCC Protein Human (HEK293 His) is the recombinant human-derived Netrin receptor DCC/DCC protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-94214 Supplier Medchemexpress LLC Supplier No. HYP78745100UG
The netrin receptor DCC guides axonal attraction in neuronal growth cones during nervous system development Interaction with UNC5 protein induces axonal repulsion Netrin receptor DCC/DCC Protein Human (HEK293 His) is the recombinant human-derived Netrin receptor DCC/DCC protein expressed by HEK293 with C-10 His labeled tag