Learn More
Medchemexpress LLC Netrin receptor DCC/ 10ug | 10ug
SDP
Catalog No. 5000195100
Encompass_Preferred
The netrin receptor DCC guides axonal attraction in neuronal growth cones during nervous system development Interaction with UNC5 protein induces axonal repulsion Netrin receptor DCC/DCC Protein Human (HEK293 His) is the recombinant human-derived Netrin receptor DCC/DCC protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-95100 Supplier Medchemexpress LLC Supplier No. HYP7874510UG
The netrin receptor DCC guides axonal attraction in neuronal growth cones during nervous system development Interaction with UNC5 protein induces axonal repulsion Netrin receptor DCC/DCC Protein Human (HEK293 His) is the recombinant human-derived Netrin receptor DCC/DCC protein expressed by HEK293 with C-10 His labeled tag