Learn More
Medchemexpress LLC Netropsin dihydrochloride | 18133-22-7 | 1 MG
SDP
Catalog No. 5000419434
Encompass_Preferred
Netropsin dihydrochloride is a small-molecule MGB (minor-groove binder) and antibiotic. It inhibits the catalytic activity of isolated topoisomerase and interferes with the stabilization of the cleavable complexes of topoisomerase II and I in nuclei. It also possesses antibacterial and antiviral activity.
  • Potential antibiotic and antiviral properties by binding to dsDNA in a non-intercalative manner.
  • Improves survival from murine endotoxaemia by attenuating NOS2 induction through interference with HMGA1 DNA binding to the core NOS2 promoter.
  • Radioprotective ability against radiation-induced damage in solutions of DNA due to high binding affinity and structural stabilization.
Catalog No. 50-004-19434 Supplier Medchemexpress LLC Supplier No. HYN6800A1MG
Netropsin dihydrochloride is a small-molecule MGB (minor-groove binder) and antibiotic. It inhibits the catalytic activity of isolated topoisomerase and interferes with the stabilization of the cleavable complexes of topoisomerase II and I in nuclei. It also possesses antibacterial and antiviral activity.
  • Potential antibiotic and antiviral properties by binding to dsDNA in a non-intercalative manner.
  • Improves survival from murine endotoxaemia by attenuating NOS2 induction through interference with HMGA1 DNA binding to the core NOS2 promoter.
  • Radioprotective ability against radiation-induced damage in solutions of DNA due to high binding affinity and structural stabilization.