Learn More
Medchemexpress LLC HY-16346 5mg Medchemexpress, Netupitant CAS:290297-26-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY163465MG
Encompass_Preferred
Medchemexpress, HY-16346 5mg Netupitant CAS:290297-26-6 Netupitant (CID-6451149) is a highly potent, selective and orally active neurokinin-1 (NK1) receptor antagonist with a Ki of 0.95 nM for hNK1 in CHO cells. Netupitant has antiemetic affect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1882