Learn More
Electron Microscopy Sciences Netwell™ Carrier Kit, 15 mm
SDP
Catalog No. 5029378
Encompass_Preferred
A set of carrier plates and handles for moving the Netwells in and out of the 6 or 12-well plates. The handle can be attached or detached to the plate at any time.
Size: 15 mm
Catalog No. 50-293-78 Supplier Electron Microscopy Sciences Supplier No. 6473500
A set of carrier plates and handles for moving the Netwells in and out of the 6 or 12-well plates. The handle can be attached or detached to the plate at any time.
Size: 15 mm