Learn More
Electron Microscopy Sciences Netwell™ Carrier Kit, 24 mm
SDP
Supplier:  Electron Microscopy Sciences 6473600
Encompass_Preferred
A modular and disposable system which can be used for the handling and processing of tissue slices in immunocytochemical staining, in 3-dimensional tissue culture, and in explant tissue culture. Home made devices are no longer necessary. These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit). A set of carrier plates and handles for moving the Netwells in and out of the 6 or 12-well plates. The handle can be attached or detached to the plate.
Size: 24 mm
Catalog No. 50-293-76