Learn More
Electron Microscopy Sciences Netwell™ Reagent Tray Black
SDP
Catalog No. 5029379
Encompass_Preferred
Black polystyrene reagent tray for Netwell Tissue Processing System, holds 150ml of solution.
Catalog No. 50-293-79 Supplier Electron Microscopy Sciences Supplier No. 6473000
Black polystyrene reagent tray for Netwell Tissue Processing System, holds 150ml of solution.