Learn More
Electron Microscopy Sciences Netwell™ Tissue Processing Set, 15 mm, 500 µm, Sterile Sterile
SDP
Catalog No. 5029368
Encompass_Preferred
A modular and disposable system which can be used for the handling and processing of tissue slices in immunocytochemical staining, in 3-dimensional tissue culture, and in explant tissue culture. Home made devices are no longer necessary. These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit). The set consists of: 12 Netwells of 15mm diameter, two reagent trays (one black and one white), one carrier kit, one carrier plate and one handle.
Size: 15 mm
Mesh: 500 µm
Sterile: Sterile
Catalog No. 50-293-68 Supplier Electron Microscopy Sciences Supplier No. 6474200
A modular and disposable system which can be used for the handling and processing of tissue slices in immunocytochemical staining, in 3-dimensional tissue culture, and in explant tissue culture. Home made devices are no longer necessary. These tissue processing wells are ready to fill all of your needs with the added convenience of being able to process up to 12 tissue specimens at one time (with one unit). The set consists of: 12 Netwells of 15mm diameter, two reagent trays (one black and one white), one carrier kit, one carrier plate and one handle.
Size: 15 mm
Mesh: 500 µm
Sterile: Sterile