Learn More
Aviva Systems Biology Corp NEU1 Antibody - middle region (ARP44286_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44286_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NEU1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human NEU1. Alternate names: NEU, SIAL1, NANHApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58771