Learn More
Medchemexpress LLC HY-106408 5mg Medchemexpress, Neu2000 CAS:640290-67-1 Purity:>98%
SDP
Catalog No. 501968342
Encompass_Preferred
Medchemexpress, HY-106408 5mg Neu2000 CAS:640290-67-1 Neu2000 is an uncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8342 Supplier Medchemexpress LLC Supplier No. HY1064085MG
Medchemexpress, HY-106408 5mg Neu2000 CAS:640290-67-1 Neu2000 is an uncompetitive N-methyl-D-aspartate (NMDA) receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.