Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neu5Ac alpha(2-3)Gal beta MP Glycoside 95.0+%, TCI America™
SDP

Catalog No. N079150MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N079150MG 50 mg
N079110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N079150MG Supplier TCI America Supplier No. N079150MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 159922-54-0
Molecular Formula C24H35NO15
Molecular Weight (g/mol) 577.536
InChI Key AECZUGNGICKBGW-GGDZMRPFSA-N
Synonym 4-Methoxyphenyl 3-O-(N-Acetyl-alpha-neuraminosyl)-beta-D-galactopyranoside, 4-Methoxyphenyl O-(5-Acetamido-3,5-dideoxy-D-glycero-alpha-D-galacto-2-nonulopyranosonic Acid)-(2-3)-beta-D-galactopyranoside, Neu5Ac alpha(2-3)Gal-beta-MP
PubChem CID 133554172
IUPAC Name (2R,4S,5S,6S)-5-acetamido-2-[(2R,3R,4R,5R,6R)-3,5-dihydroxy-2-(hydroxymethyl)-6-(4-methoxyphenoxy)oxan-4-yl]oxy-4-hydroxy-6-[(1S,2S)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
SMILES CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)OC2C(C(OC(C2O)OC3=CC=C(C=C3)OC)CO)O)O

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 577.54
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Neu5Ac alpha(2-3)Gal beta MP Glycoside
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.