Learn More
MSC Rochester Midland Corporation Biodegradable Solvent Emulsion Degreaser, NEUGENIC 4177, Food Processing
Catalog No. 502839077
Encompass
NEUGENIC 4177 is a non-petroleum, non-terpene semi-aqueous cleaner designed for a wide array of cleaning applications, particularly effective in removing oily and greasy soils from food processing and other equipment. It serves as an alternative to chlorinated solvents and CFC's in converted vapor degreasers where water rinsing is permissible.
  • Safe on food contact surfaces followed by a potable water rinse.
  • Safe on all metals.
  • Low to no odor.
  • No flash point.
  • Superior oil/water splitting ability.
  • Contains no caustic, acid.
  • Moderate foam.
  • Biodegradable.
Catalog No. 50-283-9077 Supplier MSC Supplier No. 81210205
NEUGENIC 4177 is a non-petroleum, non-terpene semi-aqueous cleaner designed for a wide array of cleaning applications, particularly effective in removing oily and greasy soils from food processing and other equipment. It serves as an alternative to chlorinated solvents and CFC's in converted vapor degreasers where water rinsing is permissible.
  • Safe on food contact surfaces followed by a potable water rinse.
  • Safe on all metals.
  • Low to no odor.
  • No flash point.
  • Superior oil/water splitting ability.
  • Contains no caustic, acid.
  • Moderate foam.
  • Biodegradable.