Learn More
ABclonal Technology NeuN Mouse mAb
SDP
Catalog No. 5000008965
Encompass_Preferred
This gene encodes a member of the RNA-binding FOX protein family which is involved in the regulation of alternative splicing of pre-mRNA The protein has an N-terminal proline-rich region an RNA recognition motif (RRM) domain and a C-terminal alanine-rich region This gene produces the neuronal nuclei (NeuN) antigen that has been widely used as a marker for post-mitotic neurons This gene has its highest expression in the central nervous system and plays a prominent role in neural tissue development and regulation of adult brain function Mutations in this gene have been associated with numerous neurological disorders Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms
Catalog No. 50-000-08965 Supplier ABclonal Technology Supplier No. A26951100UL
This gene encodes a member of the RNA-binding FOX protein family which is involved in the regulation of alternative splicing of pre-mRNA The protein has an N-terminal proline-rich region an RNA recognition motif (RRM) domain and a C-terminal alanine-rich region This gene produces the neuronal nuclei (NeuN) antigen that has been widely used as a marker for post-mitotic neurons This gene has its highest expression in the central nervous system and plays a prominent role in neural tissue development and regulation of adult brain function Mutations in this gene have been associated with numerous neurological disorders Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms