Learn More
BIO SB INC NeuN Mouse Monoclonal Antibody
Supplier:  BIO SB INC BSB 2012
Encompass
NeuN is a mouse monoclonal antibody derived from cell culture supernatant that is concentrated, dialyzed, filter sterilized and diluted in buffer pH 7.5, containing BSA and sodium azide as a preservative. Concentrated, 1:25-1:100, 1.0 mL.
Catalog No. NC1824775