Learn More
ATHENA ES Neural Basal Medium
Catalog No. 5000158126
Encompass_Preferred
Neurobasal Medium is the classic formulation used by neuroscientists for over 30 years. This neuronal cell culture medium supports long-term maintenance and maturation of pre-natal and fetal neuronal cell cultures without the need for an astrocyte feeder layer.
Catalog No. 50-001-58126 Supplier ATHENA ES Supplier No. 0468
Neurobasal Medium is the classic formulation used by neuroscientists for over 30 years. This neuronal cell culture medium supports long-term maintenance and maturation of pre-natal and fetal neuronal cell cultures without the need for an astrocyte feeder layer.