Learn More
ATCC Neural Progenitor Cells; Derived from XCL-1 GFAPp-Nanoluc-Halotag; Normal, Human (Homo sapiens)
Catalog No. 502382577
Encompass
Neural Progenitor Cells Derived from XCL-1 GFAPp-Nanoluc-Halotag is an astrocyte lineage reporter line that can be used in neuronal differentiation and drug screening.
Catalog No. 50-238-2577 Supplier ATCC Supplier No. ACS5006
Neural Progenitor Cells Derived from XCL-1 GFAPp-Nanoluc-Halotag is an astrocyte lineage reporter line that can be used in neuronal differentiation and drug screening.