Learn More
FUJIFILM Biosciences Neural supplement 91142 10ML
Catalog No. 502892205
Encompass_Preferred
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long-term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be diluted to 1X in basal medium before use. This supplement does not contain antibiotics.
Catalog No. 50-289-2205 Supplier FUJIFILM Biosciences Supplier No. 9114210ML
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long-term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be diluted to 1X in basal medium before use. This supplement does not contain antibiotics.