Learn More
Braintree Scientific NeuralGlider-Cortical Neural Implant Insertion System
SDP
Catalog No. 501955384
Encompass
NeuralGlider-Cortical Neural Implant Insertion System - Reduces insertion trauma and improves placement accuracy of penetrating neural implants NeuralGlider applies micron-scale, ultrasonic vibration to micro-wire arrays during insertion to reduce the force required to penetrate the brain surface. Reduced insertion force facilities slow (0.1 mm/s), accurate array insertions while minimizing displacement/dimpling of the cortical surface, preserving the integrity of the underlying neural tissue. Reduced insertion force and tissue dimpling yielding less insertion damage and neuron loss Integrated software for control of insertion velocity and depth Improved insertion of fixed micro-wire arrays **Sold for research purposes only and not for human use**
Catalog No. 50-195-5384 Supplier Braintree Scientific Supplier No. NG101
Add to Cart
Add to Cart
Request a Quote
NeuralGlider-Cortical Neural Implant Insertion System - Reduces insertion trauma and improves placement accuracy of penetrating neural implants NeuralGlider applies micron-scale, ultrasonic vibration to micro-wire arrays during insertion to reduce the force required to penetrate the brain surface. Reduced insertion force facilities slow (0.1 mm/s), accurate array insertions while minimizing displacement/dimpling of the cortical surface, preserving the integrity of the underlying neural tissue. Reduced insertion force and tissue dimpling yielding less insertion damage and neuron loss Integrated software for control of insertion velocity and depth Improved insertion of fixed micro-wire arrays **Sold for research purposes only and not for human use**