Learn More
New England Biolabs, Inc. α2-3,6,8 Neuraminidase - 10000 units
SDP
Catalog No. 50811833
Encompass
2-3,6,8 Neuraminidase catalyzes the hydrolysis of 2-3, 2-6, and 2-8 linked sialic acid residues from glycoproteins and oligosaccharides.
Catalog No. 50-811-833 Supplier New England Biolabs, Inc. Supplier No. P0720L
2-3,6,8 Neuraminidase catalyzes the hydrolysis of 2-3, 2-6, and 2-8 linked sialic acid residues from glycoproteins and oligosaccharides.