Learn More
Sigma Aldrich Fine Chemicals Biosciences Neuraminidase from Vibrio cholerae, 9001-67-6, MFCD00131711, 1 unit
Catalog No. NC1570700
Encompass
Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose), Synonym: Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase; Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.
Catalog No. NC1570700 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N78851UN
Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose), Synonym: Acyl-neuraminyl Hydrolase, Receptor-destroying enzyme, Sialidase; Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.