Learn More
ACROBiosystems Neuraminidase NA 100ug
Catalog No. 502534363
Encompass_Preferred
Influenza A [Wisconsin/588/2019(H1N1)] Neuraminidase (NA) Protein His Tag (NEE-V524k) is expressed from human 293 cells (HEK293). It contains AA Val 80 - Lys 469 (Accession # EPI_ISL_404527 GISAID).
Catalog No. 50-253-4363 Supplier ACROBiosystems Supplier No. NEEV524K100UG
Influenza A [Wisconsin/588/2019(H1N1)] Neuraminidase (NA) Protein His Tag (NEE-V524k) is expressed from human 293 cells (HEK293). It contains AA Val 80 - Lys 469 (Accession # EPI_ISL_404527 GISAID).