Learn More
ACROBiosystems Neuraminidase NA 100ug
Catalog No. 502534905
Encompass_Preferred
Influenza A [A/Thailand/1(KAN-1)/2004(H5N1)] Neuraminidase (NA) Protein His Tag (HA1-V5245) is expressed from human 293 cells (HEK293). It contains AA His 36 - Lys 449 (Accession # H8PF47_9INFA GISAID).
Catalog No. 50-253-4905 Supplier ACROBiosystems Supplier No. HA1V5245100UG
Influenza A [A/Thailand/1(KAN-1)/2004(H5N1)] Neuraminidase (NA) Protein His Tag (HA1-V5245) is expressed from human 293 cells (HEK293). It contains AA His 36 - Lys 449 (Accession # H8PF47_9INFA GISAID).