Learn More
ACROBiosystems Neuraminidase NA 100ug
Catalog No. 502534394
Encompass_Preferred
Influenza A [A/Darwin/9/2021 (H3N2)] Neuraminidase (NA) Protein His Tag (NE2-V5249) is expressed from human 293 cells (HEK293). It contains AA Lys 80 - Ile 469 (Accession # EPI_ISL_2233240 GISAID).
Catalog No. 50-253-4394 Supplier ACROBiosystems Supplier No. NE2V5249100UG
Influenza A [A/Darwin/9/2021 (H3N2)] Neuraminidase (NA) Protein His Tag (NE2-V5249) is expressed from human 293 cells (HEK293). It contains AA Lys 80 - Ile 469 (Accession # EPI_ISL_2233240 GISAID).