Learn More
Enzo Life Sciences Neuregulin 1-beta (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2160050
Encompass_Preferred
Alternative name: NGR beta 1, HRG1 beta, Heregulin beta1. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains 0.1% trifluoroacetic acid. Source: Produced in E. coli. A non-glycosylated protein containing 66 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4008