Learn More
Eurofins Discoverx Recombinant Human Neuregulin-1 (Heregulin β2), 50 µg. (92-1091)
Supplier:  Eurofins Discoverx 921091
Encompass
Neuregulin-1 is a glycoprotein that acts on EGFR family receptors, binding to and activating ErbB3 and ErbB4. Plays a critical role in growth and development, heart structure and function, and cardiomyocyte differentiation. Involved in development and repair of the peripheral nervous system. Provided as 50 μg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9629