Learn More
AdipoGen Neuregulin-4 (mouse) (rec.) (10 µg)
SDP
Supplier:  AdipoGen AG40B0159C010
Encompass_Preferred
Protein. Mouse Neuregulin-4 (aa 1-53) is fused at the N-terminus to a GST-tag. Source: E. coli. Endotoxin content: <0.01EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). Nrg4 is a cold induced adipokine, highly expressed in adipose tissues and enriched in brown fat. It is increased during brown adipocyte differentiation and reduced in rodent and human obesity. It promotes neurite outgrowth and protects against diet-induced insulin resistance and hepatic steatosis. This hepatic effect of Nrg4 is mediated by ErbB3 and ErbB4 signaling that negatively regulates de novo lipogenesis mediated by LXR and SREBP1c. This effect of Nrg4 on fatty liver and insulin resistance could lead to the development of Nrg4 as an effective therapeutic biological for the treatment of NAFLD and type 2 diabetes. GST-Nrg4 (aa 1-53) recombinant protein has been shown to mimic the effect of endogenous secreted Nrg4 on liver lipogenesis.
Catalog No. 50-205-3252