Learn More
Medchemexpress LLC Neuritin Human N-H 100ug
SDP
Catalog No. 5000196360
Encompass_Preferred
Recombinant human Neuritin (NRN1), expressed in E. coli with an N-terminal 6xHis tag, supplied lyophilized for research use. The protein is characterized by sequence and molecular weight (~11 kDa), high purity, and measured biological activity that supports its use in studies of neurite outgrowth and neural development.
  • Promotes neurite outgrowth and branching in neuronal cells.
  • Validated biological activity with an ED50 of approximately 17.37 ng/mL.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Recombinantly expressed in E. coli with an N-terminal 6xHis tag for detection or purification.
  • Supplied lyophilized from a buffered solution for stable storage.
  • Recommended reconstitution at or above 100 μg/mL to preserve activity.
Catalog No. 50-001-96360 Supplier Medchemexpress LLC Supplier No. HYP71908A100UG
Recombinant human Neuritin (NRN1), expressed in E. coli with an N-terminal 6xHis tag, supplied lyophilized for research use. The protein is characterized by sequence and molecular weight (~11 kDa), high purity, and measured biological activity that supports its use in studies of neurite outgrowth and neural development.
  • Promotes neurite outgrowth and branching in neuronal cells.
  • Validated biological activity with an ED50 of approximately 17.37 ng/mL.
  • High purity greater than 95% as determined by reducing SDS-PAGE.
  • Recombinantly expressed in E. coli with an N-terminal 6xHis tag for detection or purification.
  • Supplied lyophilized from a buffered solution for stable storage.
  • Recommended reconstitution at or above 100 μg/mL to preserve activity.