Learn More
Medchemexpress LLC Neuritin Human N-H 10ug
SDP
Catalog No. 5000196056
Encompass_Preferred
Recombinant human Neuritin (NRN1) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as lyophilized material for research use in neuroscience. The protein promotes neurite outgrowth and synaptic plasticity; supporting documents such as a datasheet and certificate of analysis are provided to aid experimental planning.
  • Promotes neurite outgrowth and branching in neuronal cultures.
  • Suitable for studies of synaptic plasticity and neural development.
  • Recombinant human sequence with N-terminal 6xHis tag for purification.
  • Expressed in Escherichia coli and supplied as lyophilized material.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for long-term storage.
Catalog No. 50-001-96056 Supplier Medchemexpress LLC Supplier No. HYP71908A10UG
Recombinant human Neuritin (NRN1) with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied as lyophilized material for research use in neuroscience. The protein promotes neurite outgrowth and synaptic plasticity; supporting documents such as a datasheet and certificate of analysis are provided to aid experimental planning.
  • Promotes neurite outgrowth and branching in neuronal cultures.
  • Suitable for studies of synaptic plasticity and neural development.
  • Recombinant human sequence with N-terminal 6xHis tag for purification.
  • Expressed in Escherichia coli and supplied as lyophilized material.
  • Stable when stored at -20°C; aliquot and freeze at -80°C for long-term storage.