Learn More
Medchemexpress LLC Neuritin Human N-H 50ug
SDP
Catalog No. 5000196362
Encompass_Preferred
Recombinant human Neuritin (partial length) with an N-terminal His tag, expressed in E. coli. This ~11 kDa protein promotes neurite outgrowth and synaptic plasticity and is supplied as a lyophilized powder formulated in Tris-NaCl buffer. It is intended for in vitro assays, biochemical characterization, and protein interaction studies; store frozen and follow recommended reconstitution and stabilizer guidelines for best results.
  • Expressed in E. coli and supplied with an N-terminal His tag.
  • Approximately 11 kDa partial-length protein that promotes neurite growth.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation in 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Recommended reconstitution at ≥100 μg/mL with carrier protein for stability.
Catalog No. 50-001-96362 Supplier Medchemexpress LLC Supplier No. HYP71908A50UG
Recombinant human Neuritin (partial length) with an N-terminal His tag, expressed in E. coli. This ~11 kDa protein promotes neurite outgrowth and synaptic plasticity and is supplied as a lyophilized powder formulated in Tris-NaCl buffer. It is intended for in vitro assays, biochemical characterization, and protein interaction studies; store frozen and follow recommended reconstitution and stabilizer guidelines for best results.
  • Expressed in E. coli and supplied with an N-terminal His tag.
  • Approximately 11 kDa partial-length protein that promotes neurite growth.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized formulation in 50 mM Tris-HCl, 300 mM NaCl, pH 7.4.
  • Stable when stored at -20°C; aliquot and freeze for long-term storage.
  • Recommended reconstitution at ≥100 μg/mL with carrier protein for stability.