Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neuritin Human N-H 5ug
SDP

Catalog No. 5000196363
Encompass_Preferred

Recombinant human neuritin with an N-terminal His tag supplied as a lyophilized powder for in vitro research. The protein, expressed in E. coli, is characterized for purity, endotoxin, sequence, and biological activity to support experiments on neurite outgrowth, neural development, and synaptic plasticity.

  • Lyophilized powder for long-term storage.
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg.
  • Verified biological activity with ED50 ≈ 17.37 ng/mL.
  • Includes sequence information and certificate of analysis PDFs.
  • Stable when stored at -20°C; reconstituted samples stable at 4°C for 1 week.

Catalog No. 50-001-96363 Supplier Medchemexpress LLC Supplier No. HYP71908A5UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human neuritin with an N-terminal His tag supplied as a lyophilized powder for in vitro research. The protein, expressed in E. coli, is characterized for purity, endotoxin, sequence, and biological activity to support experiments on neurite outgrowth, neural development, and synaptic plasticity.

  • Lyophilized powder for long-term storage.
  • Expressed in E. coli for efficient production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level below 1 EU/μg.
  • Verified biological activity with ED50 ≈ 17.37 ng/mL.
  • Includes sequence information and certificate of analysis PDFs.
  • Stable when stored at -20°C; reconstituted samples stable at 4°C for 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.