Learn More
Aviva Systems Biology Corp NEURL1 Antibody - N-terminal region (ARP43031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEURL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEURL. Alternate names: NEURL1, RNF67, h-neu, neu, neu-1, bA416N2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58401