Learn More
Aviva Systems Biology Corp NEURL2 Antibody - middle region (ARP52525_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52525_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NEURL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NEURL2. Alternate names: C20orf163, MGC125934, MGC125935, Ozz-E3, OZZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60762