Learn More
Vector Laboratories Neurobiotin™ Tracer for Neuronal Cell Tracing 50 mg
SDP
Catalog No. NC9313181
Encompass
NEUROBIOTIN™ Tracer is an amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for visualizing neural architecture and for the identification of gap junction coupling. NEUROBIOTIN™ Tracer can be used in many types of preparations including in vivo, whole mounts, slice preparations, or cultured cells. It can be delivered by many routes such as intracellular electrodes, microinjection, cutloading, or scrape-loading, and can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.
Catalog No. NC9313181 Supplier Vector Laboratories Supplier No. SP112050
NEUROBIOTIN™ Tracer is an amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for visualizing neural architecture and for the identification of gap junction coupling. NEUROBIOTIN™ Tracer can be used in many types of preparations including in vivo, whole mounts, slice preparations, or cultured cells. It can be delivered by many routes such as intracellular electrodes, microinjection, cutloading, or scrape-loading, and can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods.