Learn More
Biotium Neuro-DiO
Catalog No. 501964719
Encompass_Preferred
Neuro-DiO was developed as an improved alternative and replacement for the green fluorescent DiO, which has been found to be difficult to use for neurons and cell suspension due to low solubility, tendency to form aggregates and slow lateral diffusion rate. Neuro-DiO and DiO have nearly identical absorption and emission spectra, but the former has better solubility in membranes and does not form nonfluorescent aggregates, which also tend to slow down the dye diffusion rate in membranes. Also offered as ready-to-use CellBrite™ Cytoplasmic Membrane Dyes. Properties: Ex/Em = 484/501nm; Yellow-orange solid soluble in ethanol, hexane, DMSO or vegetable oil; Store at 4°C, protect from light; C67H105ClN2O5S; MW: 1086
Catalog No. 50-196-4719 Supplier Biotium Supplier No. 60015
Neuro-DiO was developed as an improved alternative and replacement for the green fluorescent DiO, which has been found to be difficult to use for neurons and cell suspension due to low solubility, tendency to form aggregates and slow lateral diffusion rate. Neuro-DiO and DiO have nearly identical absorption and emission spectra, but the former has better solubility in membranes and does not form nonfluorescent aggregates, which also tend to slow down the dye diffusion rate in membranes. Also offered as ready-to-use CellBrite™ Cytoplasmic Membrane Dyes. Properties: Ex/Em = 484/501nm; Yellow-orange solid soluble in ethanol, hexane, DMSO or vegetable oil; Store at 4°C, protect from light; C67H105ClN2O5S; MW: 1086