Learn More
Amsbio LLC Neuro2a FFPE Cell Pellet Slide (5-pack)
Catalog No. NC3588518
Encompass
SKU- 3500-3220-5pk, Size- 5 slides, Species- Mouse, Cell Line- Neuro-2a, Format- Sections, Storage- Room temperature, Shipping Temp- Ambient.
Catalog No. NC3588518 Supplier Amsbio LLC Supplier No. 350032205PK
SKU- 3500-3220-5pk, Size- 5 slides, Species- Mouse, Cell Line- Neuro-2a, Format- Sections, Storage- Room temperature, Shipping Temp- Ambient.