Learn More
ATCC Neuro-2a; Neuroblastoma; Mouse (Mus musculus)
Catalog No. 502381834
Encompass
Neuro-2a cells are mouse neuroblasts with neuronal and amoeboid stem cell morphology isolated from brain tissue. The cell line has been used for studies on the mechanism of vinblastine precipitation of microtubular protein, the kinetics of GTP binding to isolated protein, the turnover of microtubules in vivo, and the synthesis and assembly of microtubular protein. The World Organization for Animal Health (OIE) uses the cells for routine diagnosis of rabies. Use these cells to advance your neuroscience and toxicology research.
Catalog No. 50-238-1834 Supplier ATCC Supplier No. CCL131
Neuro-2a cells are mouse neuroblasts with neuronal and amoeboid stem cell morphology isolated from brain tissue. The cell line has been used for studies on the mechanism of vinblastine precipitation of microtubular protein, the kinetics of GTP binding to isolated protein, the turnover of microtubules in vivo, and the synthesis and assembly of microtubular protein. The World Organization for Animal Health (OIE) uses the cells for routine diagnosis of rabies. Use these cells to advance your neuroscience and toxicology research.