Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Neurobasal™ PhysioPlus Medium

Catalog No. A4000817201
Encompass_Preferred
Change view
Click to view available options
Without Additives:
No Phenol Red
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Without Additives
A4000817201 None
A4000817401 No Phenol Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4000817201 Supplier Gibco™ Supplier No. A4000817201
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Optimized to facilitate consistent activity, exceptional viability, and long-term maturity in a physiologically relevant system, Neurobasal PhysioPlus Medium is recommended for culturing primary and hPSC-derived neurons.

Neurobasal PhysioPlus Medium is an advanced cell culture medium for maturation and maintenance of primary and hPSC-derived neurons. The medium better replicates the central nervous system (CNS) extracellular environment and supports reliable neuronal activity that arises earlier in cultures and is sustained over long term culture durations. The medium is recommended in combination with B-27 Plus Supplement and helps provide consistent neural activity while maintaining exceptional cell viability, ensuring dependable experimental results in a physiologically relevant system.

Key Features & Benefits

Supports robust performance in functional assays

Neurobasal PhysioPlus Medium is optimized to support reliable neuronal activity and functional maturation across a wide range of neurobiology workflows, including multielectrode array (MEA) and calcium imaging applications.

  • Promotes earlier onset of spontaneous neuronal activity compared to traditional neuronal cell culture media
  • Enables a higher proportion of electrically active neurons across wells and plates
  • Supports sustained and stable network activity during long-term culture, increasing the useful experimental window of your neuronal cultures

Designed to better reflect the physiological CNS environment

Unlike conventional neuronal culture media optimized primarily for survival, Neurobasal PhysioPlus Medium was formulated to more closely mimic aspects of the extracellular environment of the central nervous system.

  • Physiologically informed formulation designed to support more in vivo-like neuronal behavior and help improve biological relevance of in vitro neuronal models
  • Optimized glucose concentration
  • Optimized neuroactive amino acid composition
  • Osmolarity aligned more closely with cerebrospinal fluid (CSF)
  • Supports electrophysiological properties, such as resting membrane potentials, spontaneous action potentials, and rheobase, more representative of native neuronal physiology, verified using whole-cell patch clamp recordings

Supports neuronal health, maturation, and long-term culture maintenance

Neurobasal PhysioPlus Medium was developed to support both neuronal function and cell health throughout extended culture durations.

  • Optimized nutrient composition supports sustained neuronal viability without compromising activity
  • Enables maintenance of healthy neuronal cultures for longer maturation and extended experimental timelines, including studies requiring more than a month in culture
  • Promotes extensive neurite outgrowth and neuronal network development

Optimized for modern neurobiology workflows

Neurobasal PhysioPlus Medium supports a broad range of neuronal cell culture applications and is recommended for use with B-27 Plus Supplement for optimal performance.

  • Compatible with both hPSC-derived neuronal models and primary rodent neurons
  • Supports applications including disease modeling, neurotoxicity studies, neuronal maturation, and functional screening
  • Available with or without phenol red to support imaging-based workflows
  • Enables researchers to generate more consistent and physiologically relevant experimental results

Built on the trusted Neurobasal legacy

Neurobasal PhysioPlus Medium builds upon the established Neurobasal platform trusted by neuroscientists for more than 30 years, delivering enhanced functional performance while maintaining the reliability and usability researchers expect from Gibco neuronal culture systems.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Neural Cells
Classification Serum-free
Form Liquid
Product Type Cell Culture Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Without Additives None
Glucose Concentration Reduced
Product Line Neurobasal
Quantity 500 mL
Shipping Condition Ambient
Tested For Endotoxin, Performance, Sterility, Osmolality, pH
Culture Type Neuronal Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.