Learn More
Vector Laboratories NEUROBIOTIN Tracer
SDP
Catalog No. 502045707
Encompass
NEUROBIOTIN Tracer (SP-1120) is an amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for visualizing neural architecture and for the identification of gap junction coupling. Can be used in many types of preparations including in vivo, whole mounts, slice preparations, or cultured cells - Can be delivered by many routes such as intracellular electrodes, microinjection, cut-loading, or scrape-loading - Can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods
Catalog No. 50-204-5707 Supplier Vector Laboratories Supplier No. SP112020
NEUROBIOTIN Tracer (SP-1120) is an amino derivative of biotin that can be used as an intracellular label for cells, particularly neurons. It is used for visualizing neural architecture and for the identification of gap junction coupling. Can be used in many types of preparations including in vivo, whole mounts, slice preparations, or cultured cells - Can be delivered by many routes such as intracellular electrodes, microinjection, cut-loading, or scrape-loading - Can be detected using avidin or streptavidin systems with either chromogenic or fluorescence visualization methods