Learn More
STEMCELL Technologies NeuroCult™ Differentiation Kit (Mouse & Rat), Size: 500 mL
SDP
Catalog No. NC0532739
Encompass
NeuroCult™ Differentiation Kit (Mouse & Rat) is a standardized medium and supplement kit for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes.
Catalog No. NC0532739 Supplier STEMCELL Technologies Supplier No. 05704
Add to Cart
Add to Cart
NeuroCult™ Differentiation Kit (Mouse & Rat) is a standardized medium and supplement kit for the differentiation of mouse and rat neural stem and progenitor cells into neurons, astrocytes, and oligodendrocytes.