Learn More
STEMCELL Technologies NeuroCult™ Proliferation Supplement (Mouse & Rat), Size: 50 mL
SDP
Supplier:  STEMCELL Technologies 05701
Encompass
NeuroCult™ Proliferation Supplement (Mouse & Rat) is a standardized, serum-free supplement for the culture of mouse and rat neural stem and progenitor cells. NeuroCult™ Proliferation Supplement (Mouse & Rat) is designed to be combined with NeuroCult™ Basal Medium (Mouse & Rat; Catalog #05700) and appropriate cytokines (not included). NeuroCult™ Proliferation Supplement (Mouse & Rat) is optimized to maintain mouse and rat neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential. This supplement is also a component of NeuroCult™ Proliferation Kit (Mouse & Rat; Catalog #05702). NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF (Catalog #78006.1) is required. When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF (Catalog #78003.1) and Heparin Solution (Catalog #07980) are also required.
Catalog No. 50-197-4352