Learn More
STEMCELL Technologies NeuroCult SM1 Without Vitamin A, Size: 10 mL
SDP
Catalog No. NC1709527
Encompass
NeuroCult™ SM1 Without Vitamin A is a serum-free culture supplement, based on the published B27 formulation, with vitamin A removed. Vitamin A, also known as retinol, induces the differentiation of neural stem cells. In combination with a basal medium of choice, NeuroCult™ SM1 Without Vitamin A can be used for primary and pluripotent stem cell (PSC)-derived neural progenitor cell expansion and in customized differentiation protocols where vitamin A is not desired.
Catalog No. NC1709527 Supplier STEMCELL Technologies Supplier No. 05731
NeuroCult™ SM1 Without Vitamin A is a serum-free culture supplement, based on the published B27 formulation, with vitamin A removed. Vitamin A, also known as retinol, induces the differentiation of neural stem cells. In combination with a basal medium of choice, NeuroCult™ SM1 Without Vitamin A can be used for primary and pluripotent stem cell (PSC)-derived neural progenitor cell expansion and in customized differentiation protocols where vitamin A is not desired.