Learn More
STEMCELL Technologies NeuroCult-XF Proliferation Medium, Size: 500 mL
SDP
Catalog No. 502390313
Encompass
NeuroCult-XF Proliferation Medium is a standardized xeno-free serum-free medium for the culture of human neural stem and progenitor cells from normal tissues or tumor samples in the neurosphere or adherent monolayer system. When supplemented with appropriate cytokines NeuroCult-XF Proliferation Medium is optimized to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal proliferation or differentiation potential.
Catalog No. 50-239-0313 Supplier STEMCELL Technologies Supplier No. 05761
NeuroCult-XF Proliferation Medium is a standardized xeno-free serum-free medium for the culture of human neural stem and progenitor cells from normal tissues or tumor samples in the neurosphere or adherent monolayer system. When supplemented with appropriate cytokines NeuroCult-XF Proliferation Medium is optimized to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal proliferation or differentiation potential.