Learn More
Aviva Systems Biology Corp NEUROD1 Antibody - N-terminal region (ARP32036_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32036_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NEUROD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEUROD1. Alternate names: BETA2, BHF-1, MODY6, NEUROD, bHLHa3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54430